missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl cyclohexylphosphonate, Thermo Scientific™

Catalog Number p-7059304
Click to view available options
Quantity:
0.5 g
2 g
10 g
This item is not returnable. View return policy
1641-61-8
C8H17O3P
192.195
MFCD00014300
OZOVYDFZXJIQPR-UHFFFAOYSA-N
dimethyl cyclohexylphosphonate, dimethylcyclohexylphosphonate, acmc-1byde, cyclohexylphosphonic acid dimethyl ester, phosphonic acid,p-cyclohexyl-,dimethyl ester, phosphonic acid,p-cyclohexyl-, dimethyl ester
12589506
dimethoxyphosphorylcyclohexane
COP(=O)(C1CCCCC1)OC
This item is not returnable. View return policy
1641-61-8
C8H17O3P
192.195
MFCD00014300
OZOVYDFZXJIQPR-UHFFFAOYSA-N
dimethyl cyclohexylphosphonate, dimethylcyclohexylphosphonate, acmc-1byde, cyclohexylphosphonic acid dimethyl ester, phosphonic acid,p-cyclohexyl-,dimethyl ester, phosphonic acid,p-cyclohexyl-, dimethyl ester
12589506
dimethoxyphosphorylcyclohexane
COP(=O)(C1CCCCC1)OC

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Boiling Point 30°C to 32°C (0.1 mmHg)
Quantity 0.5 g
Formula Weight 193.39
Physical Form Liquid
Chemical Name or Material Dimethyl cyclohexylphosphonate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'