Learn More
Dimethyl Cyclopropane-1,1-dicarboxylate, TRC
Product Code. 30383963
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
Product Code. Quantity Unit Size
30383963 500 mg 500mg
1 options
Product Code. 30383963 Supplier Toronto Research Chemicals Supplier No. TRCD477988500MG
 Restricted Item
Beräknad leverans: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Dimethyl Cyclopropane-1,1-dicarboxylate
CAS 6914-71-2
Molecular Formula C7H10O4
Quantity 500 mg
InChI Formula InChI=1S/C7H10O4/c1-10-5(8)7(3-4-7)6(9)11-2/h3-4H2,1-2H3
SMILES COC(=O)C1(CC1)C(=O)OC
IUPAC Name dimethyl cyclopropane-1,1-dicarboxylate
Molecular Weight (g/mol) 158.15
Formula Weight 158.0579