Läs mer
Dimethyl Mesaconate, TRC
Produktkod. 30371105
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30371105 100 mg
1 options
Produktkod. 30371105 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD447480100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Dimethyl Mesaconate
CAS 617-53-8
Molekylformel C7 H10 O4
Kvantitet 100 mg
Synonym (2E)-2-Methyl-2-butenedioic Acid Dimethyl Ester, (E)-2-Methyl-2-butenedioic Acid Dimethyl Ester, Mesaconic Acid Dimethyl Ester, Dimethyl Methylfumarate, NSC 226153
InChI formel InChI=1S/C7H10O4/c1-5(7(9)11-3)4-6(8)10-2/h4H,1-3H3/b5-4+
LEDER COC(=O)\C=C(/C)\C(=O)OC
IUPAC-namn dimethyl (E)-2-methylbut-2-enedioate
Molekylvikt (g/mol) 158.152
Formel vikt 158.058