missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl oxazole-4,5-dicarboxylate, 99%, Thermo Scientific™

Catalog Number p-7030521
Click to view available options
Quantity:
1 g
10 g
This item is not returnable. View return policy
72030-81-0
C7H7NO5
185.135
MFCD05863258
LYRDBSFJJDATFO-UHFFFAOYSA-N
dimethyl oxazole-4,5-dicarboxylate, 4,5-dimethyl 1,3-oxazole-4,5-dicarboxylate, methyl 5-methoxycarbonyl-1,3-oxazole-4-carboxylate
819934
dimethyl 1,3-oxazole-4,5-dicarboxylate
COC(=O)C1=C(OC=N1)C(=O)OC
This item is not returnable. View return policy
72030-81-0
C7H7NO5
185.135
MFCD05863258
LYRDBSFJJDATFO-UHFFFAOYSA-N
dimethyl oxazole-4,5-dicarboxylate, 4,5-dimethyl 1,3-oxazole-4,5-dicarboxylate, methyl 5-methoxycarbonyl-1,3-oxazole-4-carboxylate
819934
dimethyl 1,3-oxazole-4,5-dicarboxylate
COC(=O)C1=C(OC=N1)C(=O)OC

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
The teroxazole has been synthesized from dimethyl oxazole-4,5-dicarboxylate and lithiobenzyl isocyanide.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 74°C to 76°C
Boiling Point 150°C (12 mmHg)
Quantity 1 g
Formula Weight 185.14
Percent Purity 99%
Chemical Name or Material Dimethyl oxazole-4,5-dicarboxylate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'