Läs mer
Dimethyl Phosphate-d6 (Major) Sodium salt, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Dimethyl Phosphate-d6 (Major) Sodium salt |
| CAS | 157487-94-0 |
| Anteckningar om renhetsgrad | HPLC |
| Molekylformel | C10 H13 N O2 . H Cl |
| Kvantitet | 5 mg |
| Synonym | Phosphoric Acid Dimethyl Ester-d6 Sodium Salt, Dimethyl-d6 Monosodium Phosphate, Dimethyl-d6 Sodium Phosphate, Monosodium Dimethyl-d6 Phosphate, Sodium Dimethyl-d6 Phosphate |
| InChI formel | InChI=1S/C10H13NO2.ClH/c11-6-2-4-8-3-1-5-9(7-8)10(12)13;/h1,3,5,7H,2,4,6,11H2,(H,12,13);1H |
| LEDER | Cl.NCCCc1cccc(c1)C(=O)O |
| IUPAC-namn | 3-(3-aminopropyl)benzoic acid;hydrochloride |
| Molekylvikt (g/mol) | 215.677 |
| Visa mer |