Läs mer
Dimethyl Phosphorochloridate (90%), TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Dimethyl Chlorophosphate |
| CAS | 813-77-4 |
| Molekylformel | C2 H6 Cl O3 P |
| Kvantitet | 5 g |
| Synonym | Phosphorochloridic acid, dimethyl ester, Methyl phosphorochloridate (6CI,7CI), Chlorodimethoxyphosphine oxide, Chlorodimethylphosphate, Dimethoxyphosphinic chloride, Dimethoxyphosphoryl chloride, Dimethyl chlorophosphate, Dimethyl chlorophosphonate, Dimet |
| InChI formel | InChI=1S/C2H6ClO3P/c1-5-7(3,4)6-2/h1-2H3 |
| LEDER | COP(=O)(Cl)OC |
| IUPAC-namn | [chloro(methoxy)phosphoryl]oxymethane |
| Molekylvikt (g/mol) | 144.494 |
| Formel vikt | 143.974 |