Läs mer
Dimethyl phthalate-3,4,5,6-d4, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Phthalic acid, bis-methyl ester D4 |
| CAS | 93951-89-4 |
| Anteckningar om renhetsgrad | HPLC |
| Molekylformel | C10 2H4 H6 O4 |
| Kvantitet | 50 mg |
| Synonym | 1,2-Benzene-3,4,5,6-d4-dicarboxylic acid, 1,2-dimethyl ester, 1,2-Benzene-3,4,5,6-d4-dicarboxylic acid, dimethyl ester (9CI), 1,2-Dimethyl 1,2-benzene-3,4,5,6-d4-dicarboxylate, Phthalic acid-d4 dimethyl ester, Phthalic acid dimethyl ester-d4, Phthalic acid dimethyl ester D4 (phenyl-3,4,5,6-D4), Dimethyl phthalate-3,4,5,6-d4, Phthalic acid, bis-methyl ester D4, Phthalic Acid, Dimethyl Ester D4 (phenyl D4) |
| InChI formel | InChI=1S/C10H10O4/c1-13-9(11)7-5-3-4-6-8(7)10(12)14-2/h3-6H,1-2H3/i3D,4D,5D,6D |
| LEDER | [2H]c1c([2H])c([2H])c(C(=O)OC)c(C(=O)OC)c1[2H] |
| IUPAC-namn | dimethyl 3,4,5,6-tetradeuteriobenzene-1,2-dicarboxylate |
| Molekylvikt (g/mol) | 198.21 |
| Visa mer |