Läs mer
Dimethyl Succinate-d4, CDN
Specifikationer
Specifikationer
| Kemiskt namn eller material | Dimethyl Succinate-d4 |
| CAS | 30994-23-1 |
| Molekylformel | C6 D4 H6 O4 |
| Kvantitet | 1 g |
| Synonym | Butanedioic-2,2,3,3-d4 acid, 1,4-dimethyl ester (ACI), 1,4-Dimethyl butanedioate-2,2,3,3-d4 (ACI), Butanedioic-d4 acid, dimethyl ester (9 CI), Succinic-d4 acid, dimethyl ester (7 CI, 8 CI), Butanedioic-2,2,3,3-d4 acid 1,4-dimethyl ester, Succinic-d4 acid dimethyl ester, Dimethyl Succinate-d4, Succinic acid dimethyl ester-D4 (succinic-D4) |
| InChI formel | InChI=1 S/C6H10O4/c1-9-5(7)3-4-6(8)10-2/h3-4H2,1-2H3/i3D2,4D2 |
| LEDER | [2 H]C([2 H])(C(=O)OC)C([2 H])([2 H])C(=O)OC |
| IUPAC-namn | dimethyl 2,2,3,3-tetradeuteriobutanedioate |
| Molekylvikt (g/mol) | 150.1659 |
| Formel vikt | 150.083 g/mol |
| Visa mer |