Läs mer
Dimethyl sulfonazo lll (Technical Grade), TRC
Produktkod. 30375387
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30375387 250 mg
1 options
Produktkod. 30375387 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD479410250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Dimethyl sulfonazo lll (Technical Grade)
CAS 14979-11-4
Molekylformel C6 H5 N3 O
Kvantitet 250 mg
Synonym 4,5-Dihydroxy-3,6-bis[2-(4-methyl-2-sulfophenyl)diazenyl]-2,7-naphthalenedisulfonic Acid, 3,6-Bis(o-sulfo-p-methylphenylazo)chromotropic Acid, 4,5-Dihydroxy-3,6-bis[(4-methyl-2-sulfophenyl)azo]-2,7-naphthalenedisulfonic Acid, 4,5-Dihydroxy-3,6-bis[2-(4-methyl-2-sulfophenyl)diazenyl]-2,7-naphthalenedisulfonic Acid
InChI formel InChI=1S/C6H5N3O/c10-5-1-4-2-8-9-6(4)7-3-5/h1-3,10H,(H,7,8,9)
LEDER Oc1cnc2[nH]ncc2c1
IUPAC-namn 1H-pyrazolo[3,4-b]pyridin-5-ol
Molekylvikt (g/mol) 135.123
Formel vikt 135.043