missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl sulfoxide, 99.97%, MP Biomedicals™

Catalog Number p-4405544
Click to view available options
Quantity:
50 mL
250 mL
This item is not returnable. View return policy
67-68-5
C2H6OS
78.13
MFCD00002089
IAZDPXIOMUYVGZ-UHFFFAOYSA-N
dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
679
CHEBI:28262
CS(C)=O
This item is not returnable. View return policy
67-68-5
C2H6OS
78.13
MFCD00002089
IAZDPXIOMUYVGZ-UHFFFAOYSA-N
dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
679
CHEBI:28262
CS(C)=O
Color Colorless
Density 1.10g/mL
Quantity 50 mL
Physical Form Liquid To Solid
Assay Percent Range 0.9997
IUPAC Name methylsulfinylmethane
Formula Weight 78.13
Chemical Name or Material Dimethyl sulfoxide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'