Läs mer
Dimetylsulfoxid, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Dimethylsulfoxide |
| CAS | 67-68-5 |
| Molekylformel | C2 H6 O S |
| Kvantitet | 5 mL |
| Synonym | Dimethyl sulfoxide, Sulfinylbismethane, Methylsulfinylmethane, Methane, sulfinylbis- (9CI), Methyl sulfoxide (8CI), DMS 70, DMS 90, DMSO, DMSO Evol, Demasorb, Demavet, Demeso, Demsodrox, Dimethyl sulfoxide, Dimethyl sulphoxide, Dimethylsulfoxide, Dimexide, Dimexidum, Dipirartril-tropico, Dolicur, Domoso, Dromisol, Durasorb, Gamasol 90, Herpid, Hyadur, Infiltrina, Kemsol, NSC 763, Rimso 50, SQ 9453, Sclerosol, Somipront, Sulfinylbismethane, Syntexan, Methane, 1,1'-sulfinylbis- |
| InChI formel | InChI=1S/C2H6OS/c1-4(2)3/h1-2H3 |
| LEDER | CS(=O)C |
| IUPAC-namn | Metylsulfinylmetan |
| Molekylvikt (g/mol) | 78.13 |
| Formel vikt | 78.0139 |