missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Dimethylarginine Dimethylaminohydrolase 1/DDAH1 Antibody, Novus Biologicals™
Goat Polyclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NB300-916
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Dimethylarginine Dimethylaminohydrolase 1/DDAH1 Polyclonal specifically detects Dimethylarginine Dimethylaminohydrolase 1/DDAH1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Peptide ELISA.
Specifikationer
| Dimethylarginine Dimethylaminohydrolase 1/DDAH1 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| DDAH-1, DDAHdimethylargininase-1, DDAHI, Dimethylargininase-1, dimethylarginine dimethylaminohydrolase 1FLJ25539, EC 3.5.3.18, FLJ21264, N(G), N(G)-dimethylarginine dimethylaminohydrolase 1, NG, NG-dimethylarginine dimethylaminohydrolase | |
| Goat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| This antibody is expected to recognize both reported isoforms (NP_036269.1and NP_001127917.1). | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot, ELISA | |
| 0.5 mg/mL | |
| Western Blot 0.1 - 0.3 ug/ml, Peptide ELISA Detection limit 1:1000 | |
| NP_036269.1 | |
| DDAH1 | |
| Peptide with sequence TCCSVLINKKVDS corresponding to C-Terminus according to NP_036269.1, NP_001127917.1. | |
| 0.1 mg | |
| Signal Transduction | |
| 23576 | |
| Human | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
For Research Use Only
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering