Läs mer
Dimethyldithiocarbamic Acid Sodium Salt Dihydrate, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | N,N-Dimethyldithiocarbamate Sodium Dihydrate |
| CAS | 72140-17-1 |
| Molekylformel | C3 H6 N S2 . 2 H2 O . Na |
| Kvantitet | 250 mg |
| Synonym | Dimethyldithiocarbamic Acid Sodium Salt Dihydrate, Carbamodithioic acid, dimethyl-, sodium salt, dihydrate (9CI), Sodium dimethyldithiocarbamate dihydrate, N,N-Dimethyldithiocarbamate Sodium Dihydrate |
| InChI formel | InChI=1S/C3H7NS2.Na.2H2O/c1-4(2)3(5)6;;;/h1-2H3,(H,5,6);;2*1H2/q;+1;;/p-1 |
| LEDER | O.O.[Na+].CN(C)C(=S)[S-] |
| IUPAC-namn | sodium;N,N-dimethylcarbamodithioate;dihydrate |
| Molekylvikt (g/mol) | 179.237 |
| Formel vikt | 179.005 |