Learn More
Dimethyl (2-oxopropyl)phosphonate, 95%, Thermo Scientific Chemicals
Catalog Number p-3736332
765.28 SEK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
4202-14-6
C5H11O4P
166.11
MFCD00008769
UOWIYNWMROWVDG-UHFFFAOYSA-N
dimethyl 2-oxopropyl phosphonate, dimethyl acetylmethylphosphonate, dimethyl 2-oxopropylphosphonate, dimethyl acetonylphosphonate, 2-oxopropyl phosphonic acid dimethyl ester, 1-dimethoxycarbonyl acetone, acmc-209jmw, dimethoxyphosphoryl acetone
77872
1-dimethoxyphosphorylpropan-2-one
CC(=O)CP(=O)(OC)OC
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
4202-14-6
C5H11O4P
166.11
MFCD00008769
UOWIYNWMROWVDG-UHFFFAOYSA-N
dimethyl 2-oxopropyl phosphonate, dimethyl acetylmethylphosphonate, dimethyl 2-oxopropylphosphonate, dimethyl acetonylphosphonate, 2-oxopropyl phosphonic acid dimethyl ester, 1-dimethoxycarbonyl acetone, acmc-209jmw, dimethoxyphosphoryl acetone
77872
1-dimethoxyphosphorylpropan-2-one
CC(=O)CP(=O)(OC)OC
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for C-C Bond Formation (Olefination)
TRUSTED_SUSTAINABILITY
Color Colorless to Brown
Boiling Point 76°C to 79°C (3.0 mmHg)
Flash Point 93°C
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Linear Formula CH3COCH2P(O)(OCH3)2
Refractive Index 1.4380 to 1.441
Quantity 5 g
Formula Weight 166.11
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Dimethyl (2-oxopropyl)phosphonate