missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethylformamide, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™
High quality chemicals for laboratory and specialized industrial use
5837.00 SEK
Chemical Identifiers
CAS | 68-12-2 |
---|---|
Molecular Formula | C3H7NO |
Molecular Weight (g/mol) | 73.095 |
InChI Key | ZMXDDKWLCZADIW-UHFFFAOYSA-N |
Synonym | dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa |
PubChem CID | 6228 |
ChEBI | CHEBI:17741 |
IUPAC Name | N,N-dimethylformamide |
SMILES | CN(C)C=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15558544
|
JT Baker
9221-03 |
4L | Narrow Mouth Amber Glass Bottle |
5837.00 SEK
Each |
19-03-2021 |
Chemical Identifiers
68-12-2 | |
73.095 | |
dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa | |
CHEBI:17741 | |
CN(C)C=O |
C3H7NO | |
ZMXDDKWLCZADIW-UHFFFAOYSA-N | |
6228 | |
N,N-dimethylformamide |
Specifications
Dimethylformamide | |
C3H7NO | |
ZMXDDKWLCZADIW-UHFFFAOYSA-N | |
N,N-dimethylformamide | |
6228 | |
73.09 | |
Narrow Mouth Amber Glass Bottle | |
58.00°C |
68-12-2 | |
dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa | |
CN(C)C=O | |
73.095 | |
CHEBI:17741 | |
BAKER ANALYZED™ ACS Reagent | |
1L = 0.95kg | |
4L |