Läs mer
Dimethylphenylvinylsilane, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Dimethylphenylvinylsilane |
| CAS | 1125-26-4 |
| Molekylformel | C10H14Si |
| Kvantitet | 1 g |
| Synonym | (Ethenyldimethylsilyl)benzene, Dimethylvinylphenylsilane, Ethenyldimethylphenylsilane, Phenyldimethylvinylsilane, Vinyldimethylphenylsilane, Vinylphenyldimethylsilane |
| InChI formel | InChI=1S/C10H14Si/c1-4-11(2,3)10-8-6-5-7-9-10/h4-9H,1H2,2-3H3 |
| LEDER | C[Si](C)(C=C)c1ccccc1 |
| IUPAC-namn | ethenyl-dimethyl-phenylsilane |
| Molekylvikt (g/mol) | 162.3 |
| Formel vikt | 162.0865 |