Läs mer
Dimethylvinphos 100 μg/mL in Toluene, Dr. Ehrenstorfer
Produktkod. 30309310
Change view
Kvantitet:
1 ml
Produktkod. Kvantitet
30309310 1 ml
1 options
Produktkod. 30309310 Leverantör Dr. Ehrenstorfer Leverantörsnummer DREA12765000TO100
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Dimethylvinphos
CAS 2274-67-1
Analyt- eller komponentnamn Dimethylvinphos
Frakt skick Room Temperature
Molekylformel C10 H10 Cl3 O4 P
Koncentration 100 μg/mL
Certifikat och efterlevnad ISO 17034
Synonym Phosphoric acid, 2-chloro-1-(2,4-dichlorophenyl)ethenyl dimethyl ester, Phosphoric acid, 2-chloro-1-(2,4-dichlorophenyl)vinyl dimethyl ester (7CI,8CI), Benzyl alcohol, 2,4-dichloro-α-(chloromethylene)-, dimethyl phosphate (8CI), 2-Chloro-1-(2,4-dichlorophenyl)vinyl dimethyl phosphate, Chlorfenvinphos-methyl, Dimethyl-1-(2,4-dichlorophenyl)-2-chlorovinylphosphate, Dimethylvinfos, Dimethylvinphos, IPO 10, O,O-Dimethyl O-2-chloro-1-(2',4'-dichlorophenyl)-vinyl phosphate, OMS 712, Rangado, SD 8280, Shell SD 8280
InChI formel InChI=1S/C10H10Cl3O4P/c1-15-18(14,16-2)17-10(6-11)8-4-3-7(12)5-9(8)13/h3-6H,1-2H3
LEDER COP(=O)(OC)OC(=CCl)c1ccc(Cl)cc1Cl
IUPAC-namn [2-chloro-1-(2,4-dichlorophenyl)ethenyl] dimethyl phosphate
Lösningsmedel eller matris Toluene
Molekylvikt (g/mol) 331.5168
Kvantitet 1 ml
Formel vikt 329.9382 g/mol