Läs mer
Dimetofrine, TRC
Produktkod. 30368949
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30368949 100 mg
30369594 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30368949 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD479715100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 22950-29-4
Molekylformel C11 H17 N O4
Molekylvikt (g/mol) 227.26
Synonym 4-Hydroxy-3,5-dimethoxy-α-[(methylamino)methyl]benzenemethanol
IUPAC-namn 4-[1-hydroxy-2-(methylamino)ethyl]-2,6-dimethoxyphenol
LEDER CNCC(O)c1cc(OC)c(O)c(OC)c1
Specifikationer
Kemiskt namn eller material Dimetofrine
Kvantitet 100 mg
InChI formel InChI=1S/C11H17NO4/c1-12-6-8(13)7-4-9(15-2)11(14)10(5-7)16-3/h4-5,8,12-14H,6H2,1-3H3
Formel vikt 227.1158