Läs mer
Dimetotiazine Mesilate, LoGiCal
Produktkod. 30459046
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30459046 10 mg
1 options
Produktkod. 30459046 Leverantör LoGiCal Leverantörsnummer LGCFOR1591.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Dimetotiazine Mesilate
CAS 13115-40-7
Analyt- eller komponentnamn Dimetotiazine Mesilate
Frakt skick Room Temperature
Molekylformel C19 H25 N3 O2 S2 . C H4 O3 S
Certifikat och efterlevnad ISO 9001
Synonym 10H-Phenothiazine-2-sulfonamide, 10-[2-(dimethylamino)propyl]-N,N-dimethyl-, methanesulfonate (1:1), 10H-Phenothiazine-2-sulfonamide, 10-[2-(dimethylamino)propyl]-N,N-dimethyl-, monomethanesulfonate (9CI), Phenothiazine-2-sulfonamide, 10-[2-(dimethylamino)propyl]-N,N-dimethyl-, monomethanesulfonate (8CI), 10-[2-(Dimethylamino)propyl]-N,N-dimethylphenothiazine-2-sulfonamide methanesulfonate, 8599RP mesylate, Banistyl, Bonpac, Calsekin, Dimethothiazine mesylate, Dimethothiazine methanesulfonate, Fonazine mesylate, Fonazine methanesulfonate, IL 6302 mesylate, Migristene, Neomestine, Promaquid, RP 8599, RP 8599 mesylate, Yoristen
InChI formel InChI=1S/C19H25N3O2S2.CH4O3S/c1-14(20(2)3)13-22-16-8-6-7-9-18(16)25-19-11-10-15(12-17(19)22)26(23,24)21(4)5;1-5(2,3)4/h6-12,14H,13H2,1-5H3;1H3,(H,2,3,4)
Lösningstyp Neat
LEDER CC(CN1c2ccccc2Sc3ccc(cc13)S(=O)(=O)N(C)C)N(C)C.CS(=O)(=O)O
IUPAC-namn 10-[2-(dimethylamino)propyl]-N,N-dimethylphenothiazine-2-sulfonamide;methanesulfonic acid
Industry Type Clinical Testing
Molekylvikt (g/mol) 487.66
Kvantitet 10 mg
Formel vikt 487.1269 g/mol