missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimetridazole-d3, TRC
Specifications
Specifications
| Kemiskt namn eller material | Dimetridazole D3 |
| CAS | 64678-69-9 |
| Molekylformel | C5 2H3 H4 N3 O2 |
| Kvantitet | 50 mg |
| Synonym | 1H-Imidazole, 2-methyl-1-(methyl-d3)-5-nitro- (9CI), Dimetridazole-d3, Dimetridazole D3 (N-methyl D3) |
| InChI formel | InChI=1S/C5H7N3O2/c1-4-6-3-5(7(4)2)8(9)10/h3H,1-2H3/i2D3 |
| LEDER | [2H]C([2H])([2H])n1c(C)ncc1[N+](=O)[O-] |
| IUPAC-namn | 2-methyl-5-nitro-1-(trideuteriomethyl)imidazole |
| Molekylvikt (g/mol) | 144.15 |
| Formel vikt | 144.0727 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?