Läs mer
Diminazene Diaceturate, TRC
Produktkod. 17944665
Change view
Kvantitet:
50 mg
100 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17944665 50 mg
17933495 1 g
17912385 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17944665 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD47972050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 908-54-3
Molekylformel C14 H15 N7 . 2 C4 H7 N O3
Molekylvikt (g/mol) 515.52
Synonym Glycine, N-acetyl-, compd. with 4,4'-(1-triazene-1,3-diyl)bis[benzenecarboximidamide] (2:1), Benzamidine, 4,4'-diazoaminodi-, diaceturate (6CI), Glycine, N-acetyl-, compd. with 4,4'-(azoamino)dibenzamidine (7CI), Glycine, N-acetyl-, compd. with 4,4'-(diazoamino)dibenzamidine (2:1) (8CI), Benzamidine, 4,4'-(diazoamino)di-, compd. with N-acetylglycine (1:2) (8CI), Benzenecarboximidamide, 4,4'-(1-triazene-1,3-diyl)bis-, compd. with N-acetylglycine (1:2), 1,3-Bis(p-amidinophenyl)triazene bis(N-Acetylglycinate), 1,3-Bis[4-guanylphenyl]triazene diaceturate, 4,4'-(Diazoamino)dibenzamidine diaceturate, 4,4'-Diamidinodiazoaminobenzene diaceturate, Azidin, Azidine, Berenil, Beronal, Di-(4-amidinophenyl)-triazine-(N-1,3)-diaceturate, Diaminazene aceturate, Diminasan, Diminazene aceturate, Diminazene diaceturate, Diminazine aceturate, Diminazine diaceturate, Fatrybanil, Ganasag, Ganaseg, Tricip Plus, Veriben, p,p'-Diguanyldiazoaminobenzene diaceturate
IUPAC-namn 2-acetamidoacetic acid;4-[2-(4-carbamimidoylphenyl)iminohydrazinyl]benzenecarboximidamide
LEDER CC(=O)NCC(=O)O.CC(=O)NCC(=O)O.NC(=N)c1ccc(NN=Nc2ccc(cc2)C(=N)N)cc1
Specifikationer
Kemiskt namn eller material Diminazene aceturate
Anteckningar om renhetsgrad HPLC
Kvantitet 50 mg
InChI formel InChI=1S/C14H15N7.2C4H7NO3/c15-13(16)9-1-5-11(6-2-9)19-21-20-12-7-3-10(4-8-12)14(17)18;2*1-3(6)5-2-4(7)8/h1-8H,(H3,15,16)(H3,17,18)(H,19,20);2*2H2,1H3,(H,5,6)(H,7,8)
Formel vikt 515.2241
Procent renhet >95%