Läs mer
Dimorpholinophosphinyl Chloride, TRC
Produktkod. 30240642
Change view
Kvantitet:
10 g
Produktkod. Kvantitet
30240642 10 g
1 options
Produktkod. 30240642 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD47965010G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Dimorpholinophosphinyl Chloride
CAS 7264-90-6
Molekylformel C8H16ClN2O3P
Kvantitet 10 g
Synonym Bis(morpholino)phosphinyl Chloride, Bismorpholinophosphinic Chloride, Dimorpholinophosphinic Acid Chloride, Dimorpholinophosphinic Chloride, Dimorpholinophosphinic Chloride, Di-4-morpholinylphosphinic Chloride, P,P-Di-4-morpholinylphosphinic Chloride
InChI formel InChI=1S/C8H16ClN2O3P/c9-15(12,10-1-5-13-6-2-10)11-3-7-14-8-4-11/h1-8H2
LEDER ClP(=O)(N1CCOCC1)N2CCOCC2
IUPAC-namn 4-[chloro(morpholin-4-yl)phosphoryl]morpholine
Molekylvikt (g/mol) 254.65
Formel vikt 254.0587