Learn More
Lenz Laborglasintrumente Dimroth Condensers, with GL Threads
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand: Lenz Laborglasintrumente 05240311
Additional Details : Weight : 0.01000kg
Description
- Dimroth condensers acc. to DIN 12591
- For condensing vapours
- With NS socket and NS cone
- Water connection: GL 14 thread (with two threaded plastic olives)
- Made of DURAN™ tubing
Specifications
NS 14/23 | |
Duran Glass | |
250 mm |