Läs mer
Dinocap, TRC
Produktkod. 30374178
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30374178 25 mg
1 options
Produktkod. 30374178 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD48176825MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Dinocap
CAS 39300-45-3
Molekylformel C9 H5 N O4 . C8 H17 . N O2
Kvantitet 25 mg
Synonym 2-Butenoic acid 2(or 4)-isooctyl-4,6(or 2,6)-dinitrophenyl ester, Caprane, Carathane, Crotothane, DPC, DPC (pesticide), Dinocap, Karathane, Karathane 25, Karathane FN 57, Karathane LC, Mildex
InChI formel InChI=1S/C18H24N2O6/c1-4-8-17(21)26-18-14(10-7-5-6-9-13(2)3)11-15(19(22)23)12-16(18)20(24)25/h4,8,11-13H,5-7,9-10H2,1-3H3/b8-4+
LEDER [*]C(C)CCCCCC.[*]c1cc([*])c(OC(=O)C=C)c(c1)[N+](=O)[O-].[*][N+](=O)[O-]
Molekylvikt (g/mol) 350.37
Formel vikt 350.1478