Läs mer
Dioctadecyl 3,3’-Thiodipropionate, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Plastic additive 17 |
| CAS | 693-36-7 |
| Molekylformel | C42 H82 O4 S |
| Kvantitet | 50 g |
| Synonym | 3,3'-Thiobispropanoic acid 1,1'-dioctadecyl ester, 3,3'-Thiobispropanoic acid dioctadecyl ester, 3,3'-Thiodipropionic acid dioctadecyl ester, Advastab 802, Advastab PS 802, Antiox S, Antioxidant 802, Arbestab DSTDP, Carstab DSTDP, Cyanox OTDP, Cyanox STDP, DSTDP, DSTP, DSTP Yoshitomi, Dioctadecyl 3,3'-thiodipropionate, Dioctadecyl thiodipropionate, Distearyl 3,3'-thiodipropionate, Distearyl thiodipropionate, Distearyl β,β'-thiodipropionate, Distearyl β-thiodipropionate, Distearyl-3,3-thiodipropionate, Evanstab 18, Hostanox SE 2, Hostanox SE 4, Hostanox VP-SE 2, Irganox 802FD, Irganox PS 802, Irganox PS 802FD, Irganox PS 802FL, Lasumit SG, Lowinox DSTDP, Lusmit SS, NSC 65493, Naugard DSTDP, Negonox DSTP, PS 802, PS 802FL, Plastanox STDP, Plastanox STDP Antioxidant, Seenox DS, Songnox DSTDP, Stearyl 3,3'-thiodipropionate, Sumilizer TPS, Thio 1, Varox DSTDP, Yoshinox DSTDP, Yoshinox DSTP |
| InChI formel | InChI=1S/C42H82O4S/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-37-45-41(43)35-39-47-40-36-42(44)46-38-34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h3-40H2,1-2H3 |
| LEDER | CCCCCCCCCCCCCCCCCCOC(=O)CCSCCC(=O)OCCCCCCCCCCCCCCCCCC |
| IUPAC-namn | octadecyl 3-(3-octadecoxy-3-oxopropyl)sulfanylpropanoate |
| Molekylvikt (g/mol) | 683.16 |
| Formel vikt | 682.5934 |