Läs mer
Dioctanoylglycol, TRC
Produktkod. 30381285
Change view
Kvantitet:
100 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30381285 250 mg
30377956 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30381285 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD481013250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 627-86-1
Molekylformel C18H34O4
Molekylvikt (g/mol) 314.46
Synonym Dicapryloylglycol, Ethylene Glycol Dicaprylate, Ethylene Glycol Dioctanoate, Ethylenedioctanoate, NSC 97300
LEDER CCCCCCCC(=O)OCCOC(=O)CCCCCCC
Specifikationer
Kemiskt namn eller material Dioctanoylglycol
Kvantitet 250 mg
InChI formel InChI=1S/C18H34O4/c1-3-5-7-9-11-13-17(19)21-15-16-22-18(20)14-12-10-8-6-4-2/h3-16H2,1-2H3
Formel vikt 314.2457