Learn More
Thermo Scientific™ Dionex™ OnGuard™ II Ag Cartridges
Remove chloride, bromide, and iodide from sample matrices off-line to improve IC performance and facilitate trace analyses. Pretreat up to 12 samples in parallel.
Brand:  Thermo Scientific™ 057090
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 11361753
  • POA / Pack of 48
Estimated Shipment: 14-05-2024
to see stock.
This item requires a price update prior to being ordered.
Request a Price for this item here.
Description
Description
Remove chloride, bromide, and iodide from sample matrices to improve performance of ion chromatography (IC) and facilitate trace analyses. Performed prior to IC analysis, the Thermo Scientific™ Dionex™ OnGuard™ II Ag Cartridge is an off-line sample preparation technique that eliminates these matrix interferences using a silver-form, high-capacity, suffocated, cation-exchange resin. Pretreat a single sample or many samples in series. Use them singly or in series, or pretreat up to 12 samples in parallel using the optional Thermo Scientific™ Dionex™ OnGuard™ Sample Prep Station.
Dionex OnGuard II Ag Cartridge has a similar resin to and is often used in concert with the Thermo Scientific™ Dionex™ OnGuard™ II H cartridge, which removes dissolved silver and other cations. Use the Dionex OnGuard II Ag Cartridge before using the Dionex™ OnGuard II H cartridge. Also consider the Thermo Scientific™ Dionex™ OnGuard™ II Ag/H cartridge.
Solve Tough Application Problems
  • Boost sensitivity, lower baselines, and get better separations
  • Achieve reproducible trace-level determinations in concentrated matrices
  • Increase the lifetime of your analytical columns
  • Use for a wide range of matrices thanks to wide pH stability
  • Supports applications with choice of 1cc and 2.5cc formats, as well as syringe barrel format for robotic processing (custom order)
Easy-to-Use, Leak-Free
  • Leak-free, no-channeling cartridge
  • Quick, secure Luer inlet connections
Performance Guaranteed
  • Ships with a Certificate of Analysis that verifies capacity, performance and cleanliness
Pretreat 12 Samples Simultaneously
  • Use the optional Dionex OnGuard Sample Prep Station and OnGuard Sample Pretreatment Cartridges
Specifications
Specifications
Cation-exchange silver form
Dionex OnGuard Sample Prep Station
OnGuard
2.5 mL
48
Removal of Chloride, Bromide, and Iodide from Sample Matrices
Silver-Form, High-Capacity, Suffocated, Cation-Exchange Resin
OnGuard II Ag Cartridge, 2.5-cc
0 to 14
2.0 to 2.2 (1 cc); 5.0 to 5.5 (2.5 cc)