Learn More
Thermo Scientific™ Dionex™ OnGuard™ II Ba Cartridges
Description
Remove high concentrations of sulfate from sample matrices Improve ion chromatography (IC) performance and facilitate low-level ion analysis. Thermo Scientific™ Dionex™ OnGuard™ II Ba Cartridges are an off-line sample preparation technique that uses a styrene-based, sulfonic acid resin in the barium form to eliminate these matrix interferences and improve performance of ion chromatography (IC). Use them singly or in series, or pretreat up to 12 samples in parallel using the optional Thermo Scientific™ Dionex™ OnGuard™ Sample Prep Station.
Solve Tough Application Problems
- Boost sensitivity, lower baselines, and get better separations
- Achieve reproducible trace-level determinations in concentrated matrices
- Increase the lifetime of your analytical columns
- Use for a wide range of matrices thanks to wide pH stability
- Supports applications with choice of 1cc and 2.5cc formats, as well as syringe barrel format for robotic processing (custom order)
Easy-to-Use, Leak-Free
- Leak-free, no-channeling cartridge
- Quick, secure Luer inlet connections
- Ships with a Certificate of Analysis that verifies capacity, performance and cleanliness
- Use the optional Dionex OnGuard Sample Prep Station and OnGuard Sample Pretreatment Cartridges
Specifications
Specifications
| Description | Ba; 2.5 cc |
| For Use With (Application) | Removal of High Concentrations of Sulfate |
| For Use With (Equipment) | Dionex OnGuard Sample Prep Station |
| Material | Styrene-Based, Sulfonic Acid Resin in the Barium Form |
| Volume (Metric) | 2.5 mL |
| Quantity | 1 |
| Product Line | OnGuard |
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.