Läs mer
Diosmetin, MedChemExpress
Produktkod. 16491486
Encompass_Preferred
Change view
Kvantitet:
50 mg
100 mg
200 mg
Förpackningsstorlek:
200mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16491486 200 mg 200mg
16418996 100 mg -
16414767 50 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 16491486 Leverantör Medchem Express Leverantörsnummer HYN0125
MedChemExpress Diosmetin is a natural flavonoid which inhibits human CYP1A enzyme activity with an IC50 of 40 μM in HepG2 cell.
Diosmetin is a natural flavonoid which inhibits human CYP1A enzyme activity with an IC50 of 40 μM in HepG2 cell.
Specifikationer
Kemiskt namn eller material Diosmetin
CAS 520-34-3
Färg Yellow
Anteckningar om renhetsgrad Research
Molekylformel C16H12O6
Kvantitet 200 mg
Löslighetsinformation DMSO : 100 mg/mL (333.04 mM; Need ultrasonic)
LEDER O=C1C=C(C2=CC=C(OC)C(O)=C2)OC3=CC(O)=CC(O)=C13
Molekylvikt (g/mol) 300.26
Formel vikt 300.26
Procent renhet 99.8%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Cancer-programmed cell death