Läs mer
Diosmetin, TRC
Produktkod. 30376293
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30376293 100 mg
30375909 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30376293 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD485000100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Diosmetin
CAS 520-34-3
Anteckningar om renhetsgrad HPLC
Molekylformel C16 H12 O6
Kvantitet 100 mg
Synonym 4H-1-Benzopyran-4-one, 5,7-dihydroxy-2-(3-hydroxy-4-methoxyphenyl)-, Diosmetin (6CI), Flavone, 3',5,7-trihydroxy-4'-methoxy- (7CI,8CI), 3',5,7-Trihydroxy-4'-methoxyflavone, 4'-Methylluteolin, 5,7,3'-Trihydroxy-4'-methoxyflavone, 5,7-Dihydroxy-2-(3-hydroxy-4-methoxyphenyl)-4H-1-benzopyran-4-one, Diosmetine, Diosmetol, Luteolin 4'-methyl ether, Pillon, Disomin Imp. F (EP)
InChI formel InChI=1S/C16H12O6/c1-21-13-3-2-8(4-10(13)18)14-7-12(20)16-11(19)5-9(17)6-15(16)22-14/h2-7,17-19H,1H3
LEDER COc1ccc(cc1O)C2=CC(=O)c3c(O)cc(O)cc3O2
IUPAC-namn 5,7-dihydroxy-2-(3-hydroxy-4-methoxyphenyl)chromen-4-one
Molekylvikt (g/mol) 300.26
Formel vikt 300.0634