Läs mer
Diosmin EP Impurity F, Mikromol™
Produktkod. 30051905
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30051905 100 mg
30051815 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30051905 Leverantör Mikromol Leverantörsnummer MM0816.06
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Diosmin EP Impurity F
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Diosmetin
CAS 520-34-3
Analyt- eller komponentnamn Diosmetin
Frakt skick Room Temperature
Molekylformel C16 H12 O6
Certifikat och efterlevnad ISO 9001
Synonym 5,7-Dihydroxy-2-(3-hydroxy-4-methoxyphenyl)-4H-1-benzopyran-4-one (Diosmetin), 4H-1-Benzopyran-4-one, 5,7-dihydroxy-2-(3-hydroxy-4-methoxyphenyl)-, Diosmetin (6CI), Flavone, 3',5,7-trihydroxy-4'-methoxy- (7CI,8CI), 3',5,7-Trihydroxy-4'-methoxyflavone, 4'-Methylluteolin, 5,7,3'-Trihydroxy-4'-methoxyflavone, 5,7-Dihydroxy-2-(3-hydroxy-4-methoxyphenyl)-4H-1-benzopyran-4-one, Diosmetine, Diosmetol, Luteolin 4'-methyl ether, Pillon, Disomin Imp. F (EP), Diosmin Impurity F
InChI formel InChI=1S/C16H12O6/c1-21-13-3-2-8(4-10(13)18)14-7-12(20)16-11(19)5-9(17)6-15(16)22-14/h2-7,17-19H,1H3
LEDER COc1ccc(cc1O)C2=CC(=O)c3c(O)cc(O)cc3O2
IUPAC-namn 5,7-dihydroxy-2-(3-hydroxy-4-methoxyphenyl)chromen-4-one
Molekylvikt (g/mol) 300.26
Kvantitet 100 mg
Formel vikt 300.0634 g/mol