Läs mer
Diperodon Hydrochloride, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Diperodon Hydrochloride |
| CAS | 537-12-2 |
| Molekylformel | C22 H27 N3 O4 . Cl H |
| Kvantitet | 5 g |
| Synonym | 1,2-Propanediol, 3-(1-piperidinyl)-, 1,2-bis(N-phenylcarbamate), hydrochloride (1:1), 1,2-Propanediol, 3-(1-piperidinyl)-, bis(phenylcarbamate) (ester), monohydrochloride (9CI), 1,2-Propanediol, 3-piperidino-, dicarbanilate (ester), monohydrochloride (8CI), 1,2-Propanediol, 3-piperidino-, dicarbanilate, hydrochloride (6CI,7CI), 3-(1-Piperidinyl)-1,2-propanediyl bis(N-phenylcarbamate) hydrochloride, Diothane hydrochloride, Diperodon hydrochloride, NSC 76069, Proctodon |
| InChI formel | InChI=1S/C22H27N3O4.ClH/c26-21(23-18-10-4-1-5-11-18)28-17-20(16-25-14-8-3-9-15-25)29-22(27)24-19-12-6-2-7-13-19;/h1-2,4-7,10-13,20H,3,8-9,14-17H2,(H,23,26)(H,24,27);1H |
| LEDER | Cl.O=C(Nc1ccccc1)OCC(CN2CCCCC2)OC(=O)Nc3ccccc3 |
| IUPAC-namn | [2-(phenylcarbamoyloxy)-3-piperidin-1-ylpropyl] N-phenylcarbamate;hydrochloride |
| Molekylvikt (g/mol) | 433.93 |
| Formel vikt | 433.1768 |