Läs mer
Diphacinone, TRC
Produktkod. 30373501
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30373501 25 mg
1 options
Produktkod. 30373501 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD48696025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Diphacinone
CAS 82-66-6
Molekylformel C23 H16 O3
Kvantitet 25 mg
Synonym 1,3-Indandione, 2-(diphenylacetyl)- (6CI,7CI,8CI), 1H-Indene-1,3(2H)-dione, 2-(diphenylacetyl)- (9CI), 2-(2,2-Diphenylacetyl)-1H-indene-1,3(2H)-dione, 2-(Diphenylacetyl)indan-1,3-dione, Didandin, Didion, Dipaxin, Diphacin, Diphacinon, Diphacinone, Diphenacin, Diphenadion, Diphenadione, Diphenylacetylindandione, NCI 9138, NSC 9138, Oragulant, Pestoff ferret paste, Ratindan, Ratindan 1, Solvan, U 1363
InChI formel InChI=1S/C23H16O3/c24-21-17-13-7-8-14-18(17)22(25)20(21)23(26)19(15-9-3-1-4-10-15)16-11-5-2-6-12-16/h1-14,19-20H
LEDER O=C(C(c1ccccc1)c2ccccc2)C3C(=O)c4ccccc4C3=O
IUPAC-namn 2-(2,2-diphenylacetyl)indene-1,3-dione
Molekylvikt (g/mol) 340.37
Formel vikt 340.1099