Learn More
Diphenhydramine-d6 N-Oxide, TRC
Specifications
Specifications
| Kemiskt namn eller material | Diphenhydramine-d6 N-Oxide |
| CAS | 1346603-42-6 |
| Molekylformel | C17H15D6NO2 |
| Kvantitet | 1 mg |
| Synonym | 2-(Diphenylmethoxy)-N,N-(dimethyl-d6)ethanamine N-Oxide, [(Dimethyl-d6)amino]ethyl Benzhydryl Ether N-Oxide, NSC 9091-d6 |
| InChI formel | InChI=1S/C17H21NO2/c1-18(2,19)13-14-20-17(15-9-5-3-6-10-15)16-11-7-4-8-12-16/h3-12,17H,13-14H2,1-2H3/i1D3,2D3 |
| LEDER | O=N(C([2H])([2H])[2H])(C([2H])([2H])[2H])CCOC(C1=CC=CC=C1)C2=CC=CC=C2 |
| Molekylvikt (g/mol) | 277.39 |