Läs mer
Diphenhydramine N-Oxide, TRC
Produktkod. 30374968
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30374968 25 mg
1 options
Produktkod. 30374968 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD48691525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Diphenhydramine N-Oxide
CAS 3922-74-5
Anteckningar om renhetsgrad HPLC
Molekylformel C17 H21 N O2
Kvantitet 25 mg
Synonym Diphenhydramine N-Oxide, 2-(Benzhydryloxy)-N,N-dimethylethan-1-amine oxide, Ethanamine, 2-(diphenylmethoxy)-N,N-dimethyl-, N-oxide (9CI), Ethylamine, 2-(diphenylmethoxy)-N,N-dimethyl-, N-oxide (7CI,8CI), (Dimethylamino)ethyl benzhydryl ether N-oxide, NSC 9091
InChI formel InChI=1S/C17H21NO2/c1-18(2,19)13-14-20-17(15-9-5-3-6-10-15)16-11-7-4-8-12-16/h3-12,17H,13-14H2,1-2H3
LEDER C[N+](C)([O-])CCOC(c1ccccc1)c2ccccc2
IUPAC-namn 2-benzhydryloxy-N,N-dimethylethanamine oxide
Molekylvikt (g/mol) 271.35
Formel vikt 271.1572