Läs mer
Diphenhydramine, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Diphenhydramine |
| CAS | 58-73-1 |
| Anteckningar om renhetsgrad | HPLC |
| Molekylformel | C17H21NO |
| Kvantitet | 10 g |
| Synonym | Ethanamine, 2-(diphenylmethoxy)-N,N-dimethyl-, Ethylamine, 2-(diphenylmethoxy)-N,N-dimethyl- (7CI,8CI), 2-(Diphenylmethoxy)-N,N-dimethylethanamine, 2-(Benzhydryloxy)-N,N-dimethylethylamine, 2-(Diphenylmethoxy)-N,N-dimethylethylamine, Benzhydramine, DPH, Dimedrol base, Diphenhydramine, FAR 90X2, N,N-Dimethyl-2-(diphenylmethoxy)ethylamine, N-[2-(Diphenylmethoxy)ethyl]-N,N-dimethylamine, NSC 665800, O-Benzhydryldimethylaminoethanol, Probedryl, α-(2-Dimethylaminoethoxy)diphenylmethane, β-Dimethylaminoethanol diphenylmethyl ether, β-Dimethylaminoethylbenzhydrylether |
| InChI formel | InChI=1S/C17H21NO/c1-18(2)13-14-19-17(15-9-5-3-6-10-15)16-11-7-4-8-12-16/h3-12,17H,13-14H2,1-2H3 |
| LEDER | CN(C)CCOC(c1ccccc1)c2ccccc2 |
| IUPAC-namn | 2-benzhydryloxy-N,N-dimethylethanamine |
| Molekylvikt (g/mol) | 255.361 |
| Visa mer |