missing translation for 'onlineSavingsMsg'
Läs mer

Diphenidol Hydrochloride, TRC

Produktkod. 30137119
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
50 mg
1 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30137119 50 mg
30136466 1 g
30010562 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30137119 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD48694050MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 3254-89-5
Molekylformel C21 H27 N O . Cl H
Molekylvikt (g/mol) 345.91
Synonym 1-Piperidinebutanol, α,α-diphenyl-, hydrochloride (1:1), 1-Piperidinebutanol, α,α-diphenyl-, hydrochloride (6CI,7CI,8CI,9CI), Ansmin, Cefadol, Celmidol, Difenidol hydrochloride, Difenidolin, Diphenidol hydrochloride, Maniol, Mecalmin, Pineroro, SKF 478A, Satanolon, Tenesdol, Vontrol, Wansar
IUPAC-namn 1,1-diphenyl-4-piperidin-1-ylbutan-1-ol;hydrochloride
LEDER Cl.OC(CCCN1CCCCC1)(c2ccccc2)c3ccccc3

Specifikationer

Kemiskt namn eller material Difenidol Hydrochloride
Kvantitet 50 mg
InChI formel InChI=1S/C21H27NO.ClH/c23-21(19-11-4-1-5-12-19,20-13-6-2-7-14-20)15-10-18-22-16-8-3-9-17-22;/h1-2,4-7,11-14,23H,3,8-10,15-18H2;1H
Formel vikt 345.1859
Produkttitel
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.