missing translation for 'onlineSavingsMsg'
Learn More

Diphenyl N-cyanocarbonimidate, 97%, Thermo Scientific Chemicals

Catalog Number p-3736378
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
79463-77-7
C14H10N2O2
238.25
MFCD00010380
SLIKWVTWIGHFJE-UHFFFAOYSA-N
diphenyl n-cyanocarbonimidate, diphenyl cyanocarbonimidate, diphenoxymethylenecyanamide, n-cyano-diphenyl imidocarbonate, diphenylcyanocarbonimidate, cyanocarbonimidia acid diphenyl aester, n-cyanocarbonimidic acid diphenyl ester, bis phenoxy methylidenecyanamide, cyano diphenoxymethylidene amine, 3,3-diphenoxy-2-azaprop-2-enenitrile
688090
N#CN=C(OC1=CC=CC=C1)OC1=CC=CC=C1
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
79463-77-7
C14H10N2O2
238.25
MFCD00010380
SLIKWVTWIGHFJE-UHFFFAOYSA-N
diphenyl n-cyanocarbonimidate, diphenyl cyanocarbonimidate, diphenoxymethylenecyanamide, n-cyano-diphenyl imidocarbonate, diphenylcyanocarbonimidate, cyanocarbonimidia acid diphenyl aester, n-cyanocarbonimidic acid diphenyl ester, bis phenoxy methylidenecyanamide, cyano diphenoxymethylidene amine, 3,3-diphenoxy-2-azaprop-2-enenitrile
688090
N#CN=C(OC1=CC=CC=C1)OC1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 155.0°C to 160.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min
Packaging Glass bottle
Linear Formula NCN=C(OC6H5)2
Quantity 10 g
Solubility Information Solubility in water: may decompose.
IUPAC Name cyano(diphenoxymethylidene)amine
Formula Weight 238.25
Percent Purity 97%
Physical Form Fluffy Powder
Chemical Name or Material Diphenyl N-cyanocarbonimidate
Show More Show Less
compliance-icons
Product Identifier
  • Diphenyl N-cyanocarbonimidate
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'