Läs mer
Diphenyldithioperoxyanhydride, TRC
Produktkod. 30362826
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30362826 250 mg
1 options
Produktkod. 30362826 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD054750250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Diphenyldithioperoxyanhydride
CAS 5873-93-8
Molekylformel C14 H10 S4
Kvantitet 250 mg
Synonym Bis(phenylthioxomethyl) Disulfide, Bis(thiobenzoyl) Disulfide, Di(thiobenzoyl) Disulfide, Diphenyl Dithioperoxy Anhydride
InChI formel InChI=1S/C14H10S4/c15-13(11-7-3-1-4-8-11)17-18-14(16)12-9-5-2-6-10-12/h1-10H
LEDER S=C(SSC(=S)c1ccccc1)c2ccccc2
IUPAC-namn benzenecarbonothioylsulfanyl benzenecarbodithioate
Molekylvikt (g/mol) 306.489
Formel vikt 305.967