missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diphenylglyoxime, 97%, Thermo Scientific™
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Specifications
Melting Point | 238.0°C |
Assay Percent Range | 96% min. (HPLC) |
Packaging | Glass bottle |
Quantity | 25 g |
Merck Index | 15,1081 |
Solubility Information | Solubility in water: insoluble. |
Formula Weight | 240.26 |
Percent Purity | 97% |
Grade | Technical |
Chemical Name or Material | Diphenylglyoxime |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'