missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diphenyliodonium chloride, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
250 g
Description
Diphenyliodonium chloride is used as a reagent for electrophilic phenylation . It is used as a reactant for copper-catalyzed cross coupling reactions of purinesband arylation of anilines. It is involved in the preparation of acetylenic arylselenide and (arylmethylene)oxindoles. Further, it is employed in the preparation of aryl alkynes through Sonogashira coupling reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Diphenyliodonium chloride is used as a reagent for electrophilic phenylation . It is used as a reactant for copper-catalyzed cross coupling reactions of purinesband arylation of anilines. It is involved in the preparation of acetylenic arylselenide and (arylmethylene)oxindoles. Further, it is employed in the preparation of aryl alkynes through Sonogashira coupling reactions.
Solubility
Soluble in water and methanol.
Notes
Light sensitive. Incompatible with strong oxidizing agents.
Diphenyliodonium chloride is used as a reagent for electrophilic phenylation . It is used as a reactant for copper-catalyzed cross coupling reactions of purinesband arylation of anilines. It is involved in the preparation of acetylenic arylselenide and (arylmethylene)oxindoles. Further, it is employed in the preparation of aryl alkynes through Sonogashira coupling reactions.
Solubility
Soluble in water and methanol.
Notes
Light sensitive. Incompatible with strong oxidizing agents.
Specifications
Melting Point | ∼230°C (decomposition) |
Quantity | 50 g |
UN Number | UN2811 |
Beilstein | 3574977 |
Sensitivity | Light sensitive |
Merck Index | 14,7464 |
Solubility Information | Soluble in water and methanol. |
Formula Weight | 316.57 |
Percent Purity | ≥98% |
Chemical Name or Material | Diphenyliodonium chloride |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'