Learn More
Diphenyliodonium hexafluorophosphate, 97%, Thermo Scientific Chemicals
Catalog Number p-4813463
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Plastic bottle
58109-40-3
C12H10I
281.12
MFCD00061398
OZLBDYMWFAHSOQ-UHFFFAOYSA-N
diphenyliodonium hexafluorophosphate, diphenyliodanium hexafluorophosphate, diphenyliodonium hexafluorophosphate v, iodonium, diphenyl-, hexafluorophosphate 1-, iodonium, diphenyl-, hexafluorophosphate 1-1:1, diphenyliodonium pf6, dpihfp, acmc-209m2o, phi + pf6-, diphenyliodanium; hexafluoro-$l^ 5-phosphanuide
2737136
diphenyliodanium;hexafluorophosphate
[I+](C1=CC=CC=C1)C1=CC=CC=C1
58109-40-3
C12H10I
281.12
MFCD00061398
OZLBDYMWFAHSOQ-UHFFFAOYSA-N
diphenyliodonium hexafluorophosphate, diphenyliodanium hexafluorophosphate, diphenyliodonium hexafluorophosphate v, iodonium, diphenyl-, hexafluorophosphate 1-, iodonium, diphenyl-, hexafluorophosphate 1-1:1, diphenyliodonium pf6, dpihfp, acmc-209m2o, phi + pf6-, diphenyliodanium; hexafluoro-$l^ 5-phosphanuide
2737136
diphenyliodanium;hexafluorophosphate
[I+](C1=CC=CC=C1)C1=CC=CC=C1
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point 140.0°C to 144.0°C
Assay Percent Range 96% min. (HPLC)
Packaging Plastic bottle
Linear Formula (C6H5)2I+PF6-
Quantity 5 g
Solubility Information Solubility in water: .. Other solubilities: soluble in methanol
Percent Purity 97%
Chemical Name or Material Diphenyliodonium hexafluorophosphate
compliance-icons
Product Identifier
  • Diphenyliodonium hexafluorophosphate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.