Läs mer
(Diphenylphosphino)methanethiol S-Acetate, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | (Diphenylphosphino)methanethiol S-Acetate |
| CAS | 324753-11-9 |
| Molekylformel | C15 H15 O P S |
| Kvantitet | 25 mg |
| Synonym | S-[(Diphenylphosphino)methyl] ethanethioate, 1-(Diphenylphosphino)methanethiol S-Acetate, Ethanethioic Acid S-[(Diphenylphosphino)methyl] Ester |
| InChI formel | InChI=1S/C15H15OPS/c1-13(16)18-12-17(14-8-4-2-5-9-14)15-10-6-3-7-11-15/h2-11H,12H2,1H3 |
| LEDER | O=C(SCP(C=1C=CC=CC1)C=2C=CC=CC2)C |
| IUPAC-namn | S-(diphenylphosphanylmethyl) ethanethioate |
| Molekylvikt (g/mol) | 274.31776 |
| Formel vikt | 274.0581 |