Läs mer
MedChemExpress Diphenylterazine
Specifikationer
Specifikationer
| CAS | 344940-63-2 |
| Löslighet | DMSO : 27.5 mg/mL (72.86 mM∣Need ultrasonic∣DMSO can inactivate Diphenylterazine's activity) ∣DMF : 11.11 mg/mL (29.44 mM∣Need ultrasonic∣DMSO can inactivate Diphenylterazine's activity) ∣EtOH+HCl : 1 mg/mL (2.65 mM∣ultrasonic and warming and adjust pH to 2 with 1M HCl and heat to 80°C∣DMSO can inactivate Diphenylterazine's activity) ∣H2O : < 0.1 mg/mL (insoluble∣DMSO can inactivate Diphenylterazine's activity) |
| Synonym | DTZ |
| Innehåll och lagring | -80°C, protect from light |
| Kvantitet | 5 mg |
| Molekylvikt (g/mol) | 377.44 g/mol |
| Produkttyp | Dye Reagents |
| Renhet eller kvalitetsklass | 95.0% |
| Frakt skick | Room temperature |