Läs mer
Dipyridamole Tri(diethanolamine), TRC
Produktkod. 30378458
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30378458 100 mg
1 options
Produktkod. 30378458 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD492645100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Dipyridamole tri(diethanolamine)
CAS 16908-47-7
Molekylformel C23 H40 N8 O6
Kvantitet 100 mg
Synonym 2,2',2'',2''',2'''',2'''''-[[8-(Piperidin-1-yl)pyrimido[5,4-d]pyrimidine-2,4,6-triyl]trinitrilo]hexaethanol
InChI formel InChI=1S/C23H40N8O6/c32-12-6-29(7-13-33)21-19-18(24-23(27-21)31(10-16-36)11-17-37)20(28-4-2-1-3-5-28)26-22(25-19)30(8-14-34)9-15-35/h32-37H,1-17H2
LEDER OCCN(CCO)c1nc(N(CCO)CCO)c2nc(nc(N3CCCCC3)c2n1)N(CCO)CCO
IUPAC-namn 2-[[2,6-bis[bis(2-hydroxyethyl)amino]-8-piperidin-1-ylpyrimido[5,4-d]pyrimidin-4-yl]-(2-hydroxyethyl)amino]ethanol
Molekylvikt (g/mol) 524.61
Formel vikt 524.3071