missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Dipyrrolidinylthiuram Disulfide, TRC
Specifications
Specifications
| Kemiskt namn eller material | Dipyrrolidinylthiuram Disulfide |
| CAS | 496-08-2 |
| Molekylformel | C10 H16 N2 S4 |
| Kvantitet | 500 mg |
| Synonym | 1,1'-(Dithiodicarbonothioyl)bis[pyrrolidine], DPTD, Dipyrrolidylthiuram Disulfide, Lutiram, N,N,N',N'-Bis(tetramethylene)thiuram Disulfide, N,N-(1,4-Butylidene)thiuram Disulfide, Tetramethylenethiuram Disulfide |
| InChI formel | InChI=1S/C10H16N2S4/c13-9(11-5-1-2-6-11)15-16-10(14)12-7-3-4-8-12/h1-8H2 |
| LEDER | S=C(SSC(=S)N1CCCC1)N2CCCC2 |
| IUPAC-namn | pyrrolidine-1-carbothioylsulfanyl pyrrolidine-1-carbodithioate |
| Molekylvikt (g/mol) | 292.507 |
| Formel vikt | 292.02 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?