Läs mer
Dipyrrolidinylthiuram Disulfide, TRC
Produktkod. 30352390
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30352390 1 g
1 options
Produktkod. 30352390 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD2630531G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Dipyrrolidinylthiuram Disulfide
CAS 496-08-2
Molekylformel C10 H16 N2 S4
Kvantitet 1 g
Synonym 1,1'-(Dithiodicarbonothioyl)bis[pyrrolidine], DPTD, Dipyrrolidylthiuram Disulfide, Lutiram, N,N,N',N'-Bis(tetramethylene)thiuram Disulfide, N,N-(1,4-Butylidene)thiuram Disulfide, Tetramethylenethiuram Disulfide
InChI formel InChI=1S/C10H16N2S4/c13-9(11-5-1-2-6-11)15-16-10(14)12-7-3-4-8-12/h1-8H2
LEDER S=C(SSC(=S)N1CCCC1)N2CCCC2
IUPAC-namn pyrrolidine-1-carbothioylsulfanyl pyrrolidine-1-carbodithioate
Molekylvikt (g/mol) 292.507
Formel vikt 292.02