Beskrivning
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery
Specifikationer
Specifikationer
| Kemiskt namn eller material | Diquat dibromid Monohydrate |
| CAS | 6385-62-2 |
| CAS Max % | ≤ 100% |
| Linjär formel | C12H12Br2N2 · H2O |
| Beilstein | 3769318 |
| Färg | Colorless to Yellow |
| Smältpunkt | 325°C |
| Synonym | 9,10-dihydro-8a,10a-diazoniaphenanthrene dibromide; 1,1'-ethylene-2,2'-bipyridyldiylium dibromide |
| Molekylvikt (g/mol) | 362.06 g/mol |
| Kvantitet | 100 mg |