Läs mer
Diquat Dibromide, TRC
Produktkod. 30349629
Change view
Kvantitet:
500 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30349629 500 mg
30367183 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30349629 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD263085500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 85-00-7
Molekylformel C12 H12 N2 . 2 Br
Molekylvikt (g/mol) 344.05
Synonym 6,7-Dihydrodipyrido[1,2-a:2',1'-c]pyrazidiinium dibromide (6CI), 6,7-Dihydrodipyrido[1,2-a:2',1'-c]pyrazinediium dibromide (7CI), Dipyrido[1,2-a:2',1'-c]pyrazinediium, 6,7-dihydro-, dibromide (8CI,9CI), 1,1'-Dimethyl-4,4'-bipyridilium dibromide, 1,1'-Ethylene-2,2'-bipyridinium dibromide, 1,1'-Ethylene-2,2'-bipyridylium dibromide, 6,7-Dihydrodipyrido[1,2-a:2',1'-c]pyrazinium dibromide, 9,10-Dihydro-8a,10a-diazoniaphenanthrene dibromide, Diquat, Diquat dibromide, Eliminator, Eliminator (herbicide), N,N'-Ethylene-2,2'-bipyridinium dibromide, Ortho-Diquat, PP 100, PP 100 (herbicide), Reglon, Reglone, Reglone Turbo, Reglox, Reward, Weedtrine D
IUPAC-namn 6,7-dihydrodipyrido[1,2-b:1',2'-e]pyrazine-5,8-diium;dibromide
LEDER [Br-].[Br-].C1C[n+]2ccccc2c3cccc[n+]13
Specifikationer
Kemiskt namn eller material Diquat dibromide
Kvantitet 500 mg
InChI formel InChI=1S/C12H12N2.2BrH/c1-3-7-13-9-10-14-8-4-2-6-12(14)11(13)5-1;;/h1-8H,9-10H2;2*1H/q+2;;/p-2
Formel vikt 341.9367