missing translation for 'onlineSavingsMsg'
Läs mer

Diquat Dibromide, TRC

Produktkod. 30367183
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
500 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30367183 1 g
30349629 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30367183 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD2630851G

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 85-00-7
Molekylformel C12 H12 N2 . 2 Br
Molekylvikt (g/mol) 344.05
Synonym 6,7-Dihydrodipyrido[1,2-a:2',1'-c]pyrazidiinium dibromide (6CI), 6,7-Dihydrodipyrido[1,2-a:2',1'-c]pyrazinediium dibromide (7CI), Dipyrido[1,2-a:2',1'-c]pyrazinediium, 6,7-dihydro-, dibromide (8CI,9CI), 1,1'-Dimethyl-4,4'-bipyridilium dibromide, 1,1'-Ethylene-2,2'-bipyridinium dibromide, 1,1'-Ethylene-2,2'-bipyridylium dibromide, 6,7-Dihydrodipyrido[1,2-a:2',1'-c]pyrazinium dibromide, 9,10-Dihydro-8a,10a-diazoniaphenanthrene dibromide, Diquat, Diquat dibromide, Eliminator, Eliminator (herbicide), N,N'-Ethylene-2,2'-bipyridinium dibromide, Ortho-Diquat, PP 100, PP 100 (herbicide), Reglon, Reglone, Reglone Turbo, Reglox, Reward, Weedtrine D
IUPAC-namn 6,7-dihydrodipyrido[1,2-b:1',2'-e]pyrazine-5,8-diium;dibromide
LEDER [Br-].[Br-].C1C[n+]2ccccc2c3cccc[n+]13

Specifikationer

Kemiskt namn eller material Diquat dibromide
Kvantitet 1 g
InChI formel InChI=1S/C12H12N2.2BrH/c1-3-7-13-9-10-14-8-4-2-6-12(14)11(13)5-1;;/h1-8H,9-10H2;2*1H/q+2;;/p-2
Formel vikt 341.9367
Produkttitel
Välj ett problem

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.